missing translation for 'onlineSavingsMsg'
Learn More

Methyl formate, 97%, may cont. up to ca 3% methanol

Product Code. 11492538
Change view
Click to view available options
Quantity:
10,000 mL
2500 mL
500 mL
Unit Size:
10000mL
2500mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11492538 2500 mL 2500mL
11482538 500 mL 500mL
11468800 10,000 mL 10000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11492538 Supplier Thermo Scientific Alfa Aesar Supplier No. A15850.0F
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 107-31-3 | C2H4O2 | 60.05 g/mol

Methyl formate is used as a precursor to formamide, dimethylformamide and formic acid. It is used as a blowing agent and utilized for quick-drying finishes due to its high vapor pressure. It is used as a component of azeotrope to recover expensive solvents. Further, it is used as a propellant and an intermediate for optical brighteners and hydrocyanic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl formate is used as a precursor to formamide, dimethylformamide and formic acid. It is used as a blowing agent and utilized for quick-drying finishes due to its high vapor pressure. It is used as a component of azeotrope to recover expensive solvents. Further, it is used as a propellant and an intermediate for optical brighteners and hydrocyanic acid.

Solubility
Miscible with water.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-31-3
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00003291
InChI Key TZIHFWKZFHZASV-UHFFFAOYSA-N
Synonym formic acid, methyl ester, methyl methanoate, methylformiat, formic acid methyl ester, formiate de methyle, methylformiaat, mravencan methylnaty, methylformate, caswell no. 570, hcooch3
PubChem CID 7865
ChEBI CHEBI:77699
IUPAC Name methyl formate
SMILES COC=O

Specifications

Melting Point -100°C
Density 0.972
Boiling Point 32°C to 33°C
Flash Point −26°C (−14°F)
Assay Percent Range 97%
Linear Formula HCO2CH3
Refractive Index 1.344
Quantity 2500 mL
UN Number UN1243
Beilstein 1734623
Merck Index 14,6077
Solubility Information Miscible with water.
Formula Weight 60.05
Percent Purity 97%
Physical Form Liquid
Assay may cont. up to ≈3% methanol
Chemical Name or Material Methyl formate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.