missing translation for 'onlineSavingsMsg'
Learn More

α-Methyl-DL-phenylalanine, 98%, Thermo Scientific™

Product Code. 11985401 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11985401 1 g 1g
11905411 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11985401 Supplier Thermo Scientific Acros Supplier No. 444120010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 1132-26-9
Molecular Formula C10H13NO2
Molecular Weight (g/mol) 179.22
MDL Number MFCD00010512
InChI Key IRZQDMYEJPNDEN-UHFFFAOYNA-N
Synonym alpha-methyl-dl-phenylalanine, 2-amino-2-methyl-3-phenylpropionic acid, alpha-methylphenylalanine, h-dl-me phe-oh, h-alpha-me-dl-phe-oh, +/--2-amino-2-methyl-3-phenylpropionic acid, phenylalanine, alpha-methyl, 2-amino-2-methyl-3-phenyl-propanoic acid, h-me-dl-phe-oh, 2-methylphenylalanine #
PubChem CID 108055
IUPAC Name 2-amino-2-methyl-3-phenylpropanoic acid
SMILES CC(C(N)C(O)=O)C1=CC=CC=C1

Specifications

Melting Point 284°C to 286°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 179.22
Percent Purity 98%
Physical Form Powder
Chemical Name or Material α-Methyl-DL-phenylalanine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.