missing translation for 'onlineSavingsMsg'
Learn More

Methyl DL-Lactate, TRC

Product Code. 30483881
Change view
Click to view available options
Quantity:
5 mL
10 mL
Unit Size:
10mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30483881 5 mL 5mL
30478211 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30483881 Supplier Toronto Research Chemicals Supplier No. TRCM3152405ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl DL-Lactate
CAS 547-64-8
Molecular Formula C4H8O3
Quantity 5 mL
Synonym 2-Hydroxypropanoic Acid Methyl Ester, Lactic Acid Methyl Ester, (±)-Methyl 2-Hydroxypropanoate, (±)-Methyl 2-Hydroxypropionate, (±)-Methyl Lactate, DL-Methyl Lactate, Methyl 2-Hydroxypropanoate, Methyl 2-Hydroxypropionate, Methyl Lactate, Methyl alpha-Hydroxypropionate, NSC 406248
InChI Formula InChI=1S/C4H8O3/c1-3(5)4(6)7-2/h3,5H,1-2H3
SMILES COC(=O)C(C)O
IUPAC Name methyl 2-hydroxypropanoate
Molecular Weight (g/mol) 104.1
Formula Weight 104.0473
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.