missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl DL-Lactate, TRC
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Chemical Name or Material | Methyl DL-Lactate |
| CAS | 547-64-8 |
| Molecular Formula | C4H8O3 |
| Quantity | 1 mL |
| Synonym | 2-Hydroxypropanoic Acid Methyl Ester, Lactic Acid Methyl Ester, (±)-Methyl 2-Hydroxypropanoate, (±)-Methyl 2-Hydroxypropionate, (±)-Methyl Lactate, DL-Methyl Lactate, Methyl 2-Hydroxypropanoate, Methyl 2-Hydroxypropionate, Methyl Lactate, Methyl alpha-Hydroxypropionate, NSC 406248 |
| InChI Formula | InChI=1S/C4H8O3/c1-3(5)4(6)7-2/h3,5H,1-2H3 |
| SMILES | COC(=O)C(C)O |
| IUPAC Name | methyl 2-hydroxypropanoate |
| Molecular Weight (g/mol) | 104.1 |
| Formula Weight | 104.0473 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction