missing translation for 'onlineSavingsMsg'
Learn More

Methyl DL-lactate, 99%

Product Code. 10196793 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10196793 5 g 5g
10209410 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10196793 Supplier Thermo Scientific Acros Supplier No. 414480050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 547-64-8 | C4H8O3 | 104.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 547-64-8
MDL Number MFCD00066367
InChI Key LPEKGGXMPWTOCB-UHFFFAOYSA-N
Synonym methyl lactate, dl-methyl lactate, methyl dl-lactate, lactic acid methyl ester, methyl 2-hydroxypropionate, lactic acid, methyl ester, +--methyl lactate, propanoic acid, 2-hydroxy-, methyl ester, methyl alpha-hydroxypropionate, methyl-lactate
PubChem CID 11040
ChEBI CHEBI:83221
IUPAC Name methyl 2-hydroxypropanoate
SMILES CC(C(=O)OC)O

Specifications

Color Colorless to Yellow
Density 1.0900g/mL
Boiling Point 35°C (6.0 mmHg)
Flash Point 49°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C4H8O3
Linear Formula CH3CH(OH)COOCH3
Refractive Index 1.4120 to 1.414
Quantity 5 g
Merck Index 15, 6166
Specific Gravity 1.09
Specific Rotation -0.5° to +0.5° (20°C, 589nm) (neat)
Solubility Information Solubility in water: miscible,hydrolyses. Other solubilities: soluble in alcohol and ether,miscible with most common organic solvents
Molecular Weight (g/mol) 104.11
Formula Weight 104.11
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Methyl DL-lactate, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Methyl DL-lactate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.