missing translation for 'onlineSavingsMsg'
Learn More

Methyl disulfide, 99%

Product Code. 10276523 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
2.5 kg
Packaging:
Glass bottle
Unit Size:
1kg
2.5kg
250g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10276523 2.5 kg Glass bottle 2.5kg
10113673 50 g Glass bottle 50g
10235930 250 g Glass bottle 250g
10750671 1 kg Glass bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Product Code. 10276523 Supplier Thermo Scientific Acros Supplier No. 165590025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

CAS: 624-92-0 | C2H6S2 | 94.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 624-92-0
Molecular Formula C2H6S2
Molecular Weight (g/mol) 94.19
MDL Number MFCD00008561
InChI Key WQOXQRCZOLPYPM-UHFFFAOYSA-N
Synonym dimethyl disulfide, methyl disulfide, dimethyldisulfide, disulfide, dimethyl, dmds, 2,3-dithiabutane, dimethyl disulphide, methyldisulfanyl methane, methyldisulfide, methyldithiomethane
PubChem CID 12232
ChEBI CHEBI:4608
IUPAC Name (methyldisulfanyl)methane
SMILES CSSC

Specifications

Melting Point -85.0°C
Color Yellow
Density 1.0625g/mL
Boiling Point 108.0°C to 110.0°C
Flash Point 16°C
Infrared Spectrum Authentic
Assay Percent Range 1% max. (Methylmercaptan content) (GC)
Packaging Glass bottle
Linear Formula (CH3)2S2
Refractive Index 1.5243 to 1.5263
Quantity 2.5 kg
Beilstein 01, 291
Fieser 05,246; 06,217; 08,189
Specific Gravity 1.0625
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ether,ethyl alcohol,ccl4,benzene
Viscosity 0.620 mPa.s (20°C)
Formula Weight 94.19
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Methyl disulfide
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H331-Toxic if inhaled.
  • H336-May cause drowsiness or dizziness.
  • H370-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.