missing translation for 'onlineSavingsMsg'
Learn More

Methyl diphenylphosphinite, 98+%

Product Code. 11478800
Change view
Click to view available options
Quantity:
1 g
25 g
100 g
Unit Size:
100g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11478800 1 g 1g
11404603 25 g 25g
11414603 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11478800 Supplier Thermo Scientific Alfa Aesar Supplier No. A11555.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4020-99-9 | C13H13OP | 216.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4020-99-9
Molecular Formula C13H13OP
Molecular Weight (g/mol) 216.22
MDL Number MFCD00048025
InChI Key OAADXJFIBNEPLY-UHFFFAOYSA-N
Synonym methoxydiphenylphosphine, methyl diphenylphosphinite, diphenylphosphinous acid methyl ester, phosphinous acid, diphenyl-, methyl ester, methoxy diphenyl phosphane, pubchem6470, acmc-209jbw, diphenyl methoxy phosphine
PubChem CID 77636
IUPAC Name methoxy(diphenyl)phosphane
SMILES COP(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Density 1.078
Boiling Point 148°C to 150°C (6 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.604
Quantity 1 g
Beilstein 1640480
Sensitivity Air and moisture sensitive
Formula Weight 216.22
Percent Purity ≥98%
Chemical Name or Material Methyl diphenylphosphinite

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.