missing translation for 'onlineSavingsMsg'
Learn More

Methyl diphenylacetate, 98%

Product Code. 11460578
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11460578

Brand: Thermo Scientific Alfa Aesar B24694.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3469-00-9 | C15H14O2 | 226.28 g/mol

Methyl diphenylacetate is used as a chemical, organic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl diphenylacetate is used as a chemical, organic intermediate.

Solubility
Soluble in methanol.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3469-00-9
Molecular Formula C15H14O2
Molecular Weight (g/mol) 226.28
MDL Number MFCD00025869
InChI Key AORIUCNKPVHMTN-UHFFFAOYSA-N
Synonym methyl diphenylacetate, diphenylacetic acid, methyl ester, acetic acid, diphenyl-, methyl ester, benzeneacetic acid, .alpha.-phenyl-, methyl ester, methyldiphenylacetate, cbmicro_020304, acmc-209i9m, c6h5 2chcooch3, diphenylacetic acid methyl ester
PubChem CID 77019
IUPAC Name methyl 2,2-diphenylacetate
SMILES COC(=O)C(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 59°C to 61°C
Quantity 100 g
Solubility Information Soluble in methanol.
Formula Weight 226.28
Percent Purity 98%
Chemical Name or Material Methyl diphenylacetate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.