missing translation for 'onlineSavingsMsg'
Learn More

Methyl Dimethoxyacetate, TRC

Product Code. 30476128
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30476128 100 g 100g
30482411 10 g 10g
30473192 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30476128 Supplier Toronto Research Chemicals Supplier No. TRCM302860100G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89-91-8
Molecular Formula C5H10O4
Molecular Weight (g/mol) 134.13
Synonym 2,2-Dimethoxy-acetic Acid Methyl Ester, Dimethoxy-acetic Acid Methyl Ester, Glyoxylic Acid Methyl Ester 2-(Dimethyl acetal), Glyoxylic Acid Methyl Ester Dimethyl Acetal, 2,2-Dimethoxyacetic Acid Methyl Ester, Dimethoxyacetic Acid Methyl Ester, Methyl 2,2-Dimethoxyacetate, Methyl Glyoxylate Dimethyl Acetal, NSC 27791
IUPAC Name methyl 2,2-dimethoxyacetate
SMILES COC(OC)C(=O)OC

Specifications

Quantity 100 g
InChI Formula InChI=1S/C5H10O4/c1-7-4(6)5(8-2)9-3/h5H,1-3H3
Formula Weight 134.0579
Chemical Name or Material Methyl Dimethoxyacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.