missing translation for 'onlineSavingsMsg'
Learn More

Methyl dimethoxyacetate, 96%

Product Code. 11404813
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
250 g
Unit Size:
10g
250g
50g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11404813 50 g 50g
11488800 5 g 5g
11494803 10 g 10g
11414813 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11404813 Supplier Thermo Scientific Alfa Aesar Supplier No. A12171.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 89-91-8 | C5H10O4 | 134.131 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89-91-8
Molecular Formula C5H10O4
Molecular Weight (g/mol) 134.131
MDL Number MFCD00008484
InChI Key NZTCVGHPDWAALP-UHFFFAOYSA-N
Synonym methyl dimethoxyacetate, acetic acid, dimethoxy-, methyl ester, dimethoxyacetic acid methyl ester, glyoxylic acid methyl ester dimethyl acetal, acetic acid, 2,2-dimethoxy-, methyl ester, glyoxylic acid, methyl ester, 2-dimethyl acetal, zlchem 519, acmc-209r3k, ksc490c4f, methyl 2,2-dimethoxyethanoate
PubChem CID 66647
IUPAC Name methyl 2,2-dimethoxyacetate
SMILES COC(C(=O)OC)OC

Specifications

Density 1.098
Boiling Point 160°C to 163°C
Flash Point 63°C (145°F)
Refractive Index 1.405
Quantity 50 g
Beilstein 1757582
Sensitivity Moisture sensitive
Formula Weight 134.13
Percent Purity ≥97%
Chemical Name or Material Methyl dimethoxyacetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.