missing translation for 'onlineSavingsMsg'
Learn More

Methyl Diethylphosphonoacetate, TRC

Product Code. 30477695
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30477695 100 mg 100mg
30478437 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30477695 Supplier Toronto Research Chemicals Supplier No. TRCM301703100MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl Diethylphosphonoacetate
CAS 1067-74-9
Molecular Formula C7H15O5P
Quantity 100 mg
InChI Formula InChI=1S/C7H15O5P/c1-4-11-13(9,12-5-2)6-7(8)10-3/h4-6H2,1-3H3
SMILES CCOP(=O)(CC(=O)OC)OCC
IUPAC Name methyl 2-diethoxyphosphorylacetate
Molecular Weight (g/mol) 210.16
Formula Weight 210.0657
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.