missing translation for 'onlineSavingsMsg'
Learn More

Methyl diethylphosphonoacetate, 97%, Thermo Scientific Chemicals

Catalog Number 11493374
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
1067-74-9
C7H15O5P
210.166
MFCD00009081
CTSAXXHOGZNKJR-UHFFFAOYSA-N
methyl diethylphosphonoacetate, methyl 2-diethoxyphosphoryl acetate, acetic acid, diethoxyphosphinyl-, methyl ester, diethylmethylphosphonoacetate, methyl diethoxyphosphoryl acetate, diethyl carbomethoxymethylphosphonate, methyl 2-diethoxycarbonyl acetate, methyldiethylphosphonoacetate, diethyl methylphosphonoacetate, ksc489e0f
66113
methyl 2-diethoxyphosphorylacetate
CCOP(=O)(CC(=O)OC)OCC
This item is not returnable. View return policy
1067-74-9
C7H15O5P
210.166
MFCD00009081
CTSAXXHOGZNKJR-UHFFFAOYSA-N
methyl diethylphosphonoacetate, methyl 2-diethoxyphosphoryl acetate, acetic acid, diethoxyphosphinyl-, methyl ester, diethylmethylphosphonoacetate, methyl diethoxyphosphoryl acetate, diethyl carbomethoxymethylphosphonate, methyl 2-diethoxycarbonyl acetate, methyldiethylphosphonoacetate, diethyl methylphosphonoacetate, ksc489e0f
66113
methyl 2-diethoxyphosphorylacetate
CCOP(=O)(CC(=O)OC)OCC

Methyl diethylphosphonoacetate is used as a reagent in the preparation of allylic fluorides compounds and in regioselective Diels-Alder reactions. It also serves as a reactant in the preparation of substituted thiophenes and furans, which finds application in type 2 diabetes treatment. Further, it acts as a precursor in the synthesis of pyridone alkaloids and immunosuppressive cyclopentanediol derivatives. In addition to this, it is utilized in the modification of botulinum neurotoxin serotype A protease inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl diethylphosphonoacetate is used as a reagent in the preparation of allylic fluorides compounds and in regioselective Diels-Alder reactions. It also serves as a reactant in the preparation of substituted thiophenes and furans, which finds application in type 2 diabetes treatment. Further, it acts as a precursor in the synthesis of pyridone alkaloids and immunosuppressive cyclopentanediol derivatives. In addition to this, it is utilized in the modification of botulinum neurotoxin serotype A protease inhibitors.

Solubility
Miscible with tetrahydrofuran.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Density 1.145
Boiling Point 126°C to 128°C (8 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.432
Quantity 500 g
Beilstein 1782397
Solubility Information Miscible with tetrahydrofuran.
Formula Weight 210.17
Percent Purity 97%
Chemical Name or Material Methyl diethylphosphonoacetate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'