missing translation for 'onlineSavingsMsg'
Learn More

Methyl diethylphosphonoacetate, 97%

Product Code. 11483374
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11483374 100 g 100g
11473374 25 g 25g
11493374 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11483374 Supplier Thermo Scientific Alfa Aesar Supplier No. A10644.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1067-74-9 | C7H15O5P | 210.166 g/mol

Methyl diethylphosphonoacetate is used as a reagent in the preparation of allylic fluorides compounds and in regioselective Diels-Alder reactions. It also serves as a reactant in the preparation of substituted thiophenes and furans, which finds application in type 2 diabetes treatment. Further, it acts as a precursor in the synthesis of pyridone alkaloids and immunosuppressive cyclopentanediol derivatives. In addition to this, it is utilized in the modification of botulinum neurotoxin serotype A protease inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl diethylphosphonoacetate is used as a reagent in the preparation of allylic fluorides compounds and in regioselective Diels-Alder reactions. It also serves as a reactant in the preparation of substituted thiophenes and furans, which finds application in type 2 diabetes treatment. Further, it acts as a precursor in the synthesis of pyridone alkaloids and immunosuppressive cyclopentanediol derivatives. In addition to this, it is utilized in the modification of botulinum neurotoxin serotype A protease inhibitors.

Solubility
Miscible with tetrahydrofuran.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1067-74-9
Molecular Formula C7H15O5P
Molecular Weight (g/mol) 210.166
MDL Number MFCD00009081
InChI Key CTSAXXHOGZNKJR-UHFFFAOYSA-N
Synonym methyl diethylphosphonoacetate, methyl 2-diethoxyphosphoryl acetate, acetic acid, diethoxyphosphinyl-, methyl ester, diethylmethylphosphonoacetate, methyl diethoxyphosphoryl acetate, diethyl carbomethoxymethylphosphonate, methyl 2-diethoxycarbonyl acetate, methyldiethylphosphonoacetate, diethyl methylphosphonoacetate, ksc489e0f
PubChem CID 66113
IUPAC Name methyl 2-diethoxyphosphorylacetate
SMILES CCOP(=O)(CC(=O)OC)OCC

Specifications

Density 1.145
Boiling Point 126°C to 128°C (8 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.432
Quantity 100 g
Beilstein 1782397
Solubility Information Miscible with tetrahydrofuran.
Formula Weight 210.17
Percent Purity 97%
Chemical Name or Material Methyl diethylphosphonoacetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.