missing translation for 'onlineSavingsMsg'
Learn More

Methyl Diethyldithiocarbamate, TRC

Product Code. 30009755
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009755 250 mg 250mg
30005292 50 mg 50mg
30011278 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009755 Supplier Toronto Research Chemicals Supplier No. TRCM301700250MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 686-07-7
Molecular Formula C6 H13 N S2
Molecular Weight (g/mol) 163.3
Synonym Carbamic acid, diethyldithio-, methyl ester (6CI,7CI,8CI), Carbamodithioic acid, diethyl-, methyl ester (9CI), Diethyldithiocarbamate methyl ester, Methyl N,N-diethylcarbamodithioate, Methyl N,N-diethyldithiocarbamate, Methyl diethyldithiocarbamate, NSC 133269
IUPAC Name methyl N,N-diethylcarbamodithioate
SMILES CCN(CC)C(=S)SC

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C6H13NS2/c1-4-7(5-2)6(8)9-3/h4-5H2,1-3H3
Formula Weight 163.0489
Percent Purity >95%
Chemical Name or Material Methyl Diethyldithiocarbamate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.