missing translation for 'onlineSavingsMsg'
Learn More

Methyl dichloroacetate, 99%

Product Code. 11465847
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11465847 500 g 500g
11455847 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11465847 Supplier Thermo Scientific Alfa Aesar Supplier No. B24399.36
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 116-54-1 | C3H4Cl2O2 | 142.963 g/mol

Methyl dichloroacetate may be used in the diastereoselctive synthesis of bicyclic chlorocyclopropane, as difunctional initiator during atom transfer radical polymerization of methyl or n-butyl acrylate, as reagent during CrCl2-induced olefination of aldehydes. It is a reactant involved in nucleophilic substitution reactions with nitro(pentafluorosulfanyl)benzenes, azide-alkyl halide cycloaddition and atom transfer radical addition, Chemical modification of STn antigen for enhancement of synthetic carbohydrate vaccines, three component reaction of arynes with cyclic ethers and active methines, catalytic radical haloalkylation for synthesis of neodysidenin and addition reactions for syntehsis of enantiopure 3,4-diamino esters or amides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl dichloroacetate may be used in the diastereoselctive synthesis of bicyclic chlorocyclopropane, as difunctional initiator during atom transfer radical polymerization of methyl or n-butyl acrylate, as reagent during CrCl2-induced olefination of aldehydes. It is a reactant involved in nucleophilic substitution reactions with nitro(pentafluorosulfanyl)benzenes, azide-alkyl halide cycloaddition and atom transfer radical addition, Chemical modification of STn antigen for enhancement of synthetic carbohydrate vaccines, three component reaction of arynes with cyclic ethers and active methines, catalytic radical haloalkylation for synthesis of neodysidenin and addition reactions for syntehsis of enantiopure 3,4-diamino esters or amides.

Solubility
Soluble in water. (4 g/L).

Notes
Store at room temperature. Incompatible with oxidizing agents. Ensure proper ventilation.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 116-54-1
Molecular Formula C3H4Cl2O2
Molecular Weight (g/mol) 142.963
MDL Number MFCD00000843
InChI Key HKMLRUAPIDAGIE-UHFFFAOYSA-N
Synonym methyl dichloroacetate, dichloroacetic acid methyl ester, acetic acid, dichloro-, methyl ester, ccris 7740, acetic acid, 2,2-dichloro-, methyl ester, methyldichloroethanoate, acmc-1brpm, dichloroacetic acid methyl, methyl2,2-dichloroacetate, wln: gygvo1
PubChem CID 8315
IUPAC Name methyl 2,2-dichloroacetate
SMILES COC(=O)C(Cl)Cl

Specifications

Melting Point -52°C
Density 1.38
Boiling Point 142°C to 144°C
Flash Point 70°C (158°F)
Odor Pungent
Linear Formula Cl2CHCO2CH3
Refractive Index 1.442
Quantity 500 g
UN Number UN2299
Solubility Information Soluble in water. (4g/L).
Formula Weight 142.97
Percent Purity 99%
Chemical Name or Material Methyl dichloroacetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.