missing translation for 'onlineSavingsMsg'
Learn More

Methyl dichloroacetate, 99%

Product Code. 10786351 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10786351 100 mL Glass bottle 100mL
10143100 500 mL Glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10786351 Supplier Thermo Scientific Acros Supplier No. 148621000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 116-54-1 | C3H4Cl2O2 | 142.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 116-54-1
Molecular Formula C3H4Cl2O2
Molecular Weight (g/mol) 142.97
MDL Number MFCD00000843
InChI Key HKMLRUAPIDAGIE-UHFFFAOYSA-N
Synonym methyl dichloroacetate, dichloroacetic acid methyl ester, acetic acid, dichloro-, methyl ester, ccris 7740, acetic acid, 2,2-dichloro-, methyl ester, methyldichloroethanoate, acmc-1brpm, dichloroacetic acid methyl, methyl2,2-dichloroacetate, wln: gygvo1
PubChem CID 8315
IUPAC Name methyl 2,2-dichloroacetate
SMILES COC(=O)C(Cl)Cl

Specifications

Melting Point -52°C
Color Colorless
Density 1.3810g/mL
Boiling Point 141°C to 145°C
Flash Point 80°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula Cl2CHCO2CH3
Refractive Index 1.4411 to 1.4431
Quantity 100 mL
Beilstein 02, 203
Specific Gravity 1.381
Solubility Information Solubility in water: slightly soluble
Formula Weight 142.97
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Methyl dichloroacetate, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • Methyl dichloroacetate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.