missing translation for 'onlineSavingsMsg'
Learn More

Methyl deacetylasperulosidate, MedChemExpress

Product Code. 16463926
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16463926 25 mg 25mg
16461648 5 mg 5mg
16429238 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16463926 Supplier Medchem Express Supplier No. HYN1503

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Methyl deacetylasperulosidate is an iridoid and shows purgative effects in mice and lowers the blood glucose level in normal mice.

Methyl deacetylasperulosidate is an iridoid and shows purgative effects in mice and lowers the blood glucose level in normal mice.

Specifications

Chemical Name or Material Methyl deacetylasperulosidate
CAS 52613-28-2
Color White
Purity Grade Notes Research
Molecular Formula C17H24O11
Quantity 25 mg
Synonym 6α-Hydroxygeniposide Deacetylasperulosidic acid methyl ester
Solubility Information DMSO : 250 mg/mL (618.25 mM; Need ultrasonic)
SMILES O=C(C1=CO[C@@H](O[C@H]2[C@@H]([C@H]([C@@H]([C@@H](CO)O2)O)O)O)[C@@]3([H])[C@]1([H])[C@@H](O)C=C3CO)OC
Molecular Weight (g/mol) 404.37
Formula Weight 404.37
Percent Purity 98.37%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.