missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl-d3 Methanesulfonate, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | Methyl-d3 Methanesulfonate |
| CAS | 91419-94-2 |
| Molecular Formula | C2 D3 H3 O3 S |
| Quantity | 0.25 g |
| Synonym | Methan-d3-ol, 1-methanesulfonate (ACI), Methan-d3-ol, methanesulfonate (9 CI), Methanesulfonic acid, methyl-d3 ester (6 CI), Trideuteriomethyl methanesulfonate, Methyl-d3 Methanesulfonate, Methyl-D3 Methanesulfonate |
| InChI Formula | InChI=1 S/C2H6O3S/c1-5-6(2,3)4/h1-2H3/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])OS(=O)(=O)C |
| IUPAC Name | trideuteriomethyl methanesulfonate |
| Molecular Weight (g/mol) | 113.1507 |
| Formula Weight | 113.0226 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction