missing translation for 'onlineSavingsMsg'
Learn More

Methyl β-D-thiogalactoside, 98%, Thermo Scientific™

Product Code. 15552373 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15552373 250 mg 250mg
15542373 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15552373 Supplier Thermo Scientific Acros Supplier No. 459452500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 155-30-6
Molecular Formula C7H14O5S
Molecular Weight (g/mol) 210.25
InChI Key LZFNFLTVAMOOPJ-PZRMXXKTSA-N
Synonym methyl-beta-d-thiogalactoside, methyl-1-thio-beta-d-galactopyranoside, methyl 1-thio-beta-d-galactopyranoside, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-methylsulfanyl oxane-3,4,5-triol, b-d-thiomethylgalactoside, methyl b-d-thiogalactoside, galactose-derived compound 19, methyl beta-d-thio-galactopyranoside, methyl 1-thio-??-d-galactopyranoside, 1-s-methyl-1-thio-beta-d-galactopyranose
PubChem CID 10036019
IUPAC Name (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-methylsulfanyloxane-3,4,5-triol
SMILES CSC1C(C(C(C(O1)CO)O)O)O

Specifications

Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 250 mg
Solubility Information Solubility in water: soluble.
Formula Weight 210.25
Percent Purity 98%
Chemical Name or Material Methyl β-D-thiogalactoside
compliance-icons compliance-icons
Product Identifier
  • Methyl beta-D-thiogalactoside
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H370-Causes damage to organs.
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.