missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl α-D-mannopyranoside, ≥99%, Fisher BioReagents™
Description
Methyl α-D-mannopyranoside, ≥99%
- Methyl α-D-mannopyranoside is a lectin inhibitory sugar
- Competitive inhibitor of the binding of mannose by E. coli
- Intended for research use only
Specifications
Specifications
| Melting Point | 193°C to 196°C |
| Color | White |
| Loss on Drying | < 0.5% |
| Assay Percent Range | ≥99% |
| Packaging | Amber Glass |
| Quantity | 100 g |
| Specific Rotation | 76 to 85° (+ or -) |
| IUPAC Name | (2R,3S,4S,5S,6S)-2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol |
| Formula Weight | 194.2 |
| Percent Purity | ≥99% |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?