missing translation for 'onlineSavingsMsg'
Learn More

Methyl-beta-D-glucopyranoside hemihydrate, 99%, Thermo Scientific Chemicals

Product Code. 10772671
Click to view available options
25g; Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7000-27-3
Melting Point 111°C to 113°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (ELSD) (HPLC)
Packaging Glass bottle
Molecular Formula 0·5 H2O
MDL Number MFCD00149442
Quantity 25 g
Specific Rotation Condition −34.2° (20°C c=1,water)
Specific Rotation -34.2
Synonym methyl beta-d-glucoside, methyl beta-d-glucopyranoside, beta-methylglucoside, o1-methyl-glucose, beta-methyl-d-glucoside, methyl-beta-d-glucopyranoside, 1-o-methyl-beta-d-glucopyranoside, beta-d-methylglucopyranoside, methyl-beta-d-glucopyranoside hemihydrate, 2r,3s,4s,5r,6r-2-hydroxymethyl-6-methoxytetrahydro-2h-pyran-3,4,5-triol
Solubility Information (10% in water) Clear colorless
InChI Key QFDRIBVAVJNJDT-DEOAAGHXSA-N
SMILES COC1C(C(C(C(O1)CO)O)O)O.O
IUPAC Name (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol;hydrate
Molecular Weight (g/mol) 203.19
PubChem CID 75487602
Formula Weight 203.19
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Methyl-β-D-glucopyranoside hemihydrate, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'