missing translation for 'onlineSavingsMsg'
Learn More

Methyl-beta-D-galactopyranoside, 98%

Product Code. 10211283 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10211283 1 g Glass bottle 1g
10563991 5 g Glass bottle 5g
10310994 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10211283 Supplier Thermo Scientific Acros Supplier No. 314980010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1824-94-8 | C7H14O6 | 194.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1824-94-8
Molecular Formula C7H14O6
Molecular Weight (g/mol) 194.18
MDL Number MFCD00064357
InChI Key HOVAGTYPODGVJG-VOQCIKJUSA-N
Synonym methyl beta-d-galactopyranoside, methyl beta-d-galactoside, methyl galactoside, methylgalactoside, methyl beta-galactoside, galbetaome, unii-64ryd088rj, beta-d-galactopyranose methyl glycoside, beta-methyl-d-galactoside, methyl-beta-galactose
PubChem CID 94214
ChEBI CHEBI:17540
IUPAC Name (2R,3R,4S,5R,6R)-2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol
SMILES CO[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O

Specifications

Melting Point 175°C to 179°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (ELSD) (HPLC)
Packaging Glass bottle
Quantity 1 g
Specific Rotation Condition −18° (20° (c=1.5,MeOH))
Specific Rotation -18
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in methanol
Formula Weight 194.18
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Methyl-β-D-galactopyranoside
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.