missing translation for 'onlineSavingsMsg'
Learn More

Methyl Cyclopropane Carboxylate, TRC

Product Code. 30465511
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30465511 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30465511 Supplier Toronto Research Chemicals Supplier No. TRCM294710100G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cyclopropanecarboxylic Acid Methyl Ester
CAS 2868-37-3
Molecular Formula C5 H8 O2
Quantity 100 g
Synonym Cyclopropanecarboxylic acid, methyl ester, Cyclopropanecarboxylic acid methyl ester, Cyclopropanoic acid methyl ester, Methyl cyclopropanecarboxylate, Methyl cyclopropylcarboxylate
InChI Formula InChI=1S/C5H8O2/c1-7-5(6)4-2-3-4/h4H,2-3H2,1H3
SMILES COC(=O)C1CC1
IUPAC Name methyl cyclopropanecarboxylate
Molecular Weight (g/mol) 100.116
Formula Weight 100.052
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.