missing translation for 'onlineSavingsMsg'
Learn More

Methyl cyclohexanecarboxylate, 98%

Product Code. 10374002 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
50 g
Packaging:
Glass bottle
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10374002 5 g Glass bottle 5g
10317800 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10374002 Supplier Thermo Scientific Acros Supplier No. 188820050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4630-82-4 | C8H14O2 | 142.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4630-82-4
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.20
MDL Number MFCD00001458
InChI Key ZQWPRMPSCMSAJU-UHFFFAOYSA-N
Synonym cyclohexanecarboxylic acid, methyl ester, methyl cyclohexanoate, methyl cyclohexylcarboxylate, cyclohexanecarboxylic acid methyl ester, hexahydrobenzoic acid methyl ester, methyl cyclohexylformate, methyl hexahydrobenzoate, methyl cyclohexane carboxylate, fema no. 3568, methyl ester of cyclohexanecarboxylic acid
PubChem CID 20748
IUPAC Name methyl cyclohexanecarboxylate
SMILES COC(=O)C1CCCCC1

Specifications

Color Colorless to Yellow
Density 0.9950g/mL
Boiling Point 183°C
Flash Point 60°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula C6H11CO2CH3
Refractive Index 1.4420 to 1.444
Quantity 5 g
Specific Gravity 0.995
Solubility Information Solubility in water: almost insoluble in water
Formula Weight 142.2
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Methyl cyclohexanecarboxylate, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Methyl cyclohexanecarboxylate
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.