missing translation for 'onlineSavingsMsg'
Learn More

Methyl-β-cyclodextrin, MedChemExpress

Product Code. 16439906
Encompass_Preferred
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
50 g
Unit Size:
10g
1g
25g
50g
5g
This item is not returnable. View return policy

Product Code. 16439906

Brand: Medchem Express HY101461

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Methyl-β-cyclodextrin (Methyl-beta-cyclodextrin) is a cyclic heptasaccharide used to deliver hydrophobic drugs based on its property of solubilizing non-polar substances. Methyl-β-cyclodextrin is also extensively used as a cholesterol-depleting reagent. Methyl-β-cyclodextrin strongly reduces clathrin-dependent endocytosis.

Methyl-β-cyclodextrin (Methyl-beta-cyclodextrin) is a cyclic heptasaccharide used to deliver hydrophobic drugs based on its property of solubilizing non-polar substances. Methyl-β-cyclodextrin is also extensively used as a cholesterol-depleting reagent. Methyl-β-cyclodextrin strongly reduces clathrin-dependent endocytosis.

Specifications

Chemical Name or Material Methyl-β-cyclodextrin
CAS 128446-36-6
Color White
Purity Grade Notes Research
Quantity 50 g
Synonym Methyl-beta-cyclodextrin
Solubility Information DMSO : ≥ 100 mg/mL ∣H2O : ≥ 50 mg/mL
SMILES CO[C@@H]1[C@H](O[R])[C@H](O[R])[C@@H](C)C(CO[R])O1.[7].[R=H or -CH3]
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.