missing translation for 'onlineSavingsMsg'
Learn More

Methyl Cyanoacetate, TRC

Product Code. 30464829
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30464829 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30464829 Supplier Toronto Research Chemicals Supplier No. TRCM294995100G
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl Cyanoacetate
CAS 105-34-0
Molecular Formula C4H5NO2
Quantity 100 g
Synonym 2-Cyano-acetic Acid Methyl Ester, Cyano-acetic Acid Methyl Ester, (Methoxycarbonyl)acetonitrile, 2-Cyanoacetic Acid Methyl Ester, Cyanoacetic Acid Methyl Ester, Methyl 2-Cyanoacetate, Methyl Cyanoethanoate, Methyl alpha-Cyanoacetate, NSC 3113, NSC 68507
InChI Formula InChI=1S/C4H5NO2/c1-7-4(6)2-3-5/h2H2,1H3
SMILES COC(=O)CC#N
IUPAC Name methyl 2-cyanoacetate
Molecular Weight (g/mol) 99.09
Formula Weight 99.032
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.