missing translation for 'onlineSavingsMsg'
Learn More

Methyl cyanoacetate, 99%

Product Code. 11492997
Change view
Click to view available options
Quantity:
250 g
500 g
1000 g
Unit Size:
1000g
250g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11492997 1000 g 1000g
11403007 250 g 250g
11408810 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11492997 Supplier Thermo Scientific Alfa Aesar Supplier No. A15555.0B
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 105-34-0 | C4H5NO2 | 99.09 g/mol

Methyl cyanoacetate is an intermediate in the preparation of α-cyanoacrylate, malononitrile, 1,1-cyclohexanediacetic acid dye. It is used as UV absorber or stabiliser, intermediate for pharmaceuticals. It may be used in the synthesis of various 1,2,5-tricarbonyl compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl cyanoacetate is an intermediate in the preparation of α-cyanoacrylate, malononitrile, 1,1-cyclohexanediacetic acid dye. It is used as UV absorber or stabiliser, intermediate for pharmaceuticals. It may be used in the synthesis of various 1,2,5-tricarbonyl compounds.

Solubility
Soluble in water 117 g/L.

Notes
Light sensitive. Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105-34-0
Molecular Formula C4H5NO2
Molecular Weight (g/mol) 99.09
MDL Number MFCD00001939
InChI Key ANGDWNBGPBMQHW-UHFFFAOYSA-N
Synonym methyl cyanoacetate, cyanoacetic acid methyl ester, methyl cyanoethanoate, acetic acid, cyano-, methyl ester, usaf kf-22, methylester kyseliny kyanoctove, methylcyanoacetate, malonic methyl ester nitrile, acetic acid, 2-cyano-, methyl ester, methyl cyano acetate
PubChem CID 7747
ChEBI CHEBI:51928
IUPAC Name methyl 2-cyanoacetate
SMILES COC(=O)CC#N

Specifications

Melting Point -23°C
Density 1.127
Boiling Point 204°C
Flash Point 110°C (230°F)
Odor Mild
Refractive Index 1.4185
Quantity 1000 g
Beilstein 773945
Solubility Information Soluble in water 117g/L.
Formula Weight 99.09
Percent Purity 99%
Chemical Name or Material Methyl cyanoacetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.