missing translation for 'onlineSavingsMsg'
Learn More

Methyl cis-11-Eicosenoate, TRC

Product Code. 30134596
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134596 250 mg 250mg
30009529 100 mg 100mg
30009862 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134596 Supplier Toronto Research Chemicals Supplier No. TRCM303020250MG
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2390-09-2
Molecular Formula C21 H40 O2
Molecular Weight (g/mol) 324.54
Synonym (11Z)-11-Eicosenoic Acid Methyl Ester, 11-Eicosenoic acid, methyl ester, (Z)- (8CI), Methyl (Z)-11-eicosenoate, Methyl 11(Z)-eicosenoate, Methyl 11-cis-eicosenoate, cis-11-Eicosenoic acid methyl ester
IUPAC Name methyl (Z)-icos-11-enoate
SMILES CCCCCCCC\C=C/CCCCCCCCCC(=O)OC

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C21H40O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21(22)23-2/h10-11H,3-9,12-20H2,1-2H3/b11-10-
Formula Weight 324.3028
Percent Purity >95%
Chemical Name or Material Gondolic Acid Methyl Ester
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.