missing translation for 'onlineSavingsMsg'
Learn More

Methyl cinnamate, predominantly trans, 99%

Product Code. 11468546
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11468546 100 g 100g
11478546 500 g 500g
11488546 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11468546 Supplier Thermo Scientific Alfa Aesar Supplier No. A15975.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 103-26-4 | C10H10O2 | 162.188 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Methyl cinnamate was used to inhibit monophenolase and diphenolase activity of mushroom tyrosinase and it also has antimicrobial ability. It is mainly is used in the flavor and perfume industries. It is known to attract males of various orchid bees, such as Aglae caerulea.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-26-4
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.188
MDL Number MFCD00008458
InChI Key CCRCUPLGCSFEDV-BQYQJAHWSA-N
Synonym methyl cinnamate, methyl trans-cinnamate, methyl e-cinnamate, cinnamic acid methyl ester, methyl cinnamylate, methyl 3-phenylacrylate, methyl e-3-phenylprop-2-enoate, methyl 3-phenylpropenoate, trans-cinnamic acid methyl ester, trans-methyl cinnamate
PubChem CID 637520
ChEBI CHEBI:6857
IUPAC Name methyl (E)-3-phenylprop-2-enoate
SMILES COC(=O)C=CC1=CC=CC=C1

Specifications

Melting Point 34°C to 37°C
Density 1.042
Boiling Point 262°C to 264°C
Flash Point 123°C (253°F)
Assay Percent Range 99%
Refractive Index 1.577
Quantity 100 g
Beilstein 386468
Merck Index 14,2299
Solubility Information Insoluble in water.
Formula Weight 162.19
Percent Purity 99%
Chemical Name or Material Methyl Cinnamate, predominantly trans
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.