missing translation for 'onlineSavingsMsg'
Learn More

Methyl cinnamate, 98%

Product Code. p-161250 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
500 g
Packaging:
Glass bottle
Unit Size:
500g
5g
This item is not returnable. View return policy

Product Code. 10783251

Brand: Thermo Scientific Acros 279760050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 103-26-4 | C10H10O2 | 162.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-26-4
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.19
MDL Number MFCD00008458
InChI Key CCRCUPLGCSFEDV-BQYQJAHWSA-N
Synonym methyl cinnamate, methyl trans-cinnamate, methyl e-cinnamate, cinnamic acid methyl ester, methyl cinnamylate, methyl 3-phenylacrylate, methyl e-3-phenylprop-2-enoate, methyl 3-phenylpropenoate, trans-cinnamic acid methyl ester, trans-methyl cinnamate
PubChem CID 637520
ChEBI CHEBI:6857
IUPAC Name methyl (E)-3-phenylprop-2-enoate
SMILES COC(=O)C=CC1=CC=CC=C1

Specifications

Acidity 1mg KOH/g max.
Melting Point 33.5°C to 36°C
Color White to Yellow
Boiling Point 260°C to 262°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH=CHCO2CH3
Quantity 5 g
Beilstein 09, 581
Merck Index 15, 2299
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether,glycerol,most fixed,oils and mineral oil
Formula Weight 162.19
Percent Purity 98%
Physical Form Fused Crystalline Mass
Chemical Name or Material Methyl cinnamate, 98%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.