missing translation for 'onlineSavingsMsg'
Learn More

Methyl Cholate, Mikromol™

Product Code. 30051818
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
25 mg
missing translation for 'unitSize':
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity missing translation for 'unitSize'
30051818 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30051818 missing translation for 'mfr' Mikromol missing translation for 'supplierNo' MM0835.120025
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Methyl Cholate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cholic Acid Methyl Ester
CAS 1448-36-8
Analyte or Component Names Cholic Acid Methyl Ester
Shipping Condition Room Temperature
Molecular Formula C25 H42 O5
Certifications/Compliance ISO 9001
Synonym Methyl Cholate, Cholan-24-oic acid, 3,7,12-trihydroxy-, methyl ester, (3α,5β,7α,12α)-, Cholic acid, methyl ester (6CI,7CI,8CI), Methyl 3α,7α,12α-trihydroxy-5β-cholan-24-ate, Methyl 3α,7α,12α-trihydroxy-5β-cholan-24-oate, Methyl 3α,7α,12α-trihydroxy-5β-cholanoate, Methyl cholate, NSC 126794
InChI Formula InChI=1S/C25H42O5/c1-14(5-8-22(29)30-4)17-6-7-18-23-19(13-21(28)25(17,18)3)24(2)10-9-16(26)11-15(24)12-20(23)27/h14-21,23,26-28H,5-13H2,1-4H3/t14-,15+,16-,17-,18+,19+,20-,21+,23+,24+,25-/m1/s1
SMILES COC(=O)CC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3[C@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C
IUPAC Name methyl (4R)-4-[(3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoate
Molecular Weight (g/mol) 422.6
Quantity 25 mg
Formula Weight 422.3032 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.