missing translation for 'onlineSavingsMsg'
Learn More

Methyl Chlorooxoacetate, TRC

Product Code. 30467575
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30467575 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30467575 Supplier Toronto Research Chemicals Supplier No. TRCM29540550G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl Chlorooxoacetate
CAS 5781-53-3
Molecular Formula C3H3ClO3
Quantity 50 g
Synonym 2-Chloro-2-oxoacetic Acid Methyl Ester, Chloroglyoxylic Acid Methyl Ester, Methoxalyl Chloride, Methyl (Chlorocarbonyl)formate, Methyl (Chloroformyl)formate, Methyl 2-Chloro-2-oxoacetate, Methyl Chloroglyoxylate, Methyl Chlorooxalate, Methyl Oxalyl Chloride, Monomethyl Oxalate Chloride, Monomethyl Oxalate Monochloride, Oxalic Acid Methyl Ester Chloride, Oxalic Acid Monomethyl Ester Chloride
InChI Formula InChI=1S/C3H3ClO3/c1-7-3(6)2(4)5/h1H3
SMILES ClC(C(OC)=O)=O
IUPAC Name methyl 2-chloro-2-oxoacetate
Molecular Weight (g/mol) 122.51
Formula Weight 121.9771
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.