missing translation for 'onlineSavingsMsg'
Learn More

Methyl chloroformate, 99%

Product Code. 10327200 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
Unit Size:
100mL
2.5 L
500mL
5mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10327200 500 mL Glass bottle 500mL
10317200 5 mL Glass bottle 5mL
10261592 100 mL Glass bottle 100mL
10125783 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 10327200 Supplier Thermo Scientific Acros Supplier No. 126535000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 79-22-1 | C2H3ClO2 | 94.49 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-22-1 , 75-44-5 , 67-56-1
Molecular Formula C2H3ClO2
Molecular Weight (g/mol) 94.49
MDL Number MFCD00000639
InChI Key XMJHPCRAQCTCFT-UHFFFAOYSA-N
Synonym methyl chloroformate, methyl chlorocarbonate, chloroformic acid methyl ester, methoxycarbonyl chloride, carbonochloridic acid, methyl ester, methyl chloridocarbonate, chloro methoxy methanone, metilcloroformiato, k-stoff, methylchloorformiaat
PubChem CID 6586
IUPAC Name methyl carbonochloridate
SMILES COC(Cl)=O

Specifications

Melting Point -61.0°C
Color Colorless to Yellow
Density 1.2230g/mL
Boiling Point 70.0°C to 72.0°C
Flash Point 5°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula ClCO2CH3
Refractive Index 1.3860 to 1.3880
Quantity 500 mL
Beilstein 03, 9
Fieser 06,376; 07,236; 09,306; 11,336
Merck Index 15, 6114
Specific Gravity 1.223
Solubility Information Solubility in water: hydrolysis. Other solubilities: miscible with alcohol, methanol, benzene,, chloroform and ether
Viscosity 0.48 mPa.s (20°C)
Formula Weight 94.5
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Methyl chloroformate, 97%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methyl chloroformate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H300-Fatal if swallowed.
  • H312-Harmful in contact with skin.
  • H290-May be corrosive to metals.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.