missing translation for 'onlineSavingsMsg'
Learn More

Methyl chloroacetate, 99+%

Product Code. 11441870
Change view
Click to view available options
Quantity:
250 g
2500 g
Unit Size:
2500g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11441870 2500 g 2500g
11451870 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11441870 Supplier Thermo Scientific Alfa Aesar Supplier No. A10325.0E
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 96-34-4 | C3H5ClO2 | 108.521 g/mol

Methyl chloroacetate is used as a solvent and chemical intermediate. It acts as a precursor in the preparation of (carboxymethyl) trimethylammonium chloride esters. Further, it is used in the preparation of octakis-(carbethoxymethoxy)calix[8]arene. It is employed as an extraction solvent during the separation of neutral compounds. In addition to this, it is used in the synthesis of dimethyl carbonate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl chloroacetate is used as a solvent and chemical intermediate. It acts as a precursor in the preparation of (carboxymethyl) trimethylammonium chloride esters. Further, it is used in the preparation of octakis-(carbethoxymethoxy)calix[8]arene. It is employed as an extraction solvent during the separation of neutral compounds. In addition to this, it is used in the synthesis of dimethyl carbonate.

Solubility
Miscible with organic solvents, alcohol, ether, acetone and benzene. Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-34-4
Molecular Formula C3H5ClO2
Molecular Weight (g/mol) 108.521
MDL Number MFCD00000931
InChI Key QABLOFMHHSOFRJ-UHFFFAOYSA-N
Synonym methyl chloroacetate, chloroacetic acid methyl ester, methyl monochloroacetate, methyl chloroethanoate, acetic acid, chloro-, methyl ester, methyl monochloracetate, methyl alpha-chloroacetate, methylchloroacetate, unii-450vsb182i, ccris 7749
PubChem CID 7295
IUPAC Name methyl 2-chloroacetate
SMILES COC(=O)CCl

Specifications

Melting Point -32°C
Density 1.236
Boiling Point 128°C to 130°C
Flash Point 51°C (123°F)
Refractive Index 1.422
Quantity 2500 g
UN Number UN2295
Beilstein 506255
Merck Index 14,6042
Solubility Information Miscible with organic solvents,alcohol,ether,acetone and benzene. Slightly miscible with water.
Formula Weight 108.52
Percent Purity ≥99%
Chemical Name or Material Methyl chloroacetate
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methyl chloroacetate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H310-Fatal in contact with skin.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H335-May cause respiratory irritation.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Methyl chloroacetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.