missing translation for 'onlineSavingsMsg'
Learn More

Methyl chloroacetate, 98%

Product Code. 10376690 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
Unit Size:
1 L
10 L
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10376690 2.5 L Glass bottle 2.5 L
10326600 1 L Glass bottle 1 L
10036043 10 L Plastic drum 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 10376690 Supplier Thermo Scientific Acros Supplier No. 148630025
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 96-34-4 | C3H5ClO2 | 108.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-34-4
Molecular Formula C3H5ClO2
Molecular Weight (g/mol) 108.52
MDL Number MFCD00000931
InChI Key QABLOFMHHSOFRJ-UHFFFAOYSA-N
Synonym methyl chloroacetate, chloroacetic acid methyl ester, methyl monochloroacetate, methyl chloroethanoate, acetic acid, chloro-, methyl ester, methyl monochloracetate, methyl alpha-chloroacetate, methylchloroacetate, unii-450vsb182i, ccris 7749
PubChem CID 7295
IUPAC Name methyl 2-chloroacetate
SMILES COC(=O)CCl

Specifications

Melting Point -33.0°C
Color Colorless
Density 1.2300g/mL
Boiling Point 130.0°C to 132.0°C
Flash Point 47°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula ClCH2CO2CH3
Refractive Index 1.4210 to 1.4230
Quantity 2.5 L
Beilstein 02,197
Merck Index 15,6113
Specific Gravity 1.23
Solubility Information Solubility in water: 28g/L (20°C). Other solubilities: soluble in methanol,ethanol,chloroform,,toluene,acetone,heptane,hexane,cyclohexane,,xylene and ether
Viscosity 1.23 mPa.s (20°C)
Formula Weight 108.52
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Methyl chloroacetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.