missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Cellulose, Viscosity of 2% aqueous solution at 25°C: 4,000 centipoises, MP Biomedicals
Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 g
Unit Size:
100g
Specifications
Specifications
| CAS | 9004-67-5 |
| Molecular Formula | C20H38O11 |
| Quantity | 100 g |
| InChI Key | YLGXILFCIXHCMC-JHGZEJCSSA-N |
| SMILES | COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC |
| IUPAC Name | (5R)-2,3,4-trimethoxy-6-(methoxymethyl)-5-[(2S)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane |
| Molecular Weight (g/mol) | 454.513 |
| PubChem CID | 51063134 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction