missing translation for 'onlineSavingsMsg'
Learn More

Methyl Cellulose, viscosity of 2% aqueous solution (25°C) 15cps, MP Biomedicals

Product Code. 11497772 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
Unit Size:
100g
1kg
250g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11497772 100 g 100g
11407782 250 g 250g
11417782 500 g 500g
11427782 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11497772 Supplier MP Biomedicals Supplier No. 0215549280
 Restricted Item
Estimated Shipment: 12-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 9004-67-5
Molecular Formula C20H38O11
Molecular Weight (g/mol) 454.513
InChI Key YLGXILFCIXHCMC-JHGZEJCSSA-N
Synonym Cellulose methyl ether
PubChem CID 51063134
IUPAC Name (5R)-2,3,4-trimethoxy-6-(methoxymethyl)-5-[(2S)-3,4,5-trimethoxy-6-(methoxymethyl)oxan-2-yl]oxyoxane
SMILES COCC1C(C(C(C(O1)OC2C(OC(C(C2OC)OC)OC)COC)OC)OC)OC

Specifications

Color White
Physical Form Powder
Quantity 100 g
Chemical Name or Material Methyl Cellulose, Viscosity of 2% aqueous solution at 25°C: 15cps
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.