missing translation for 'onlineSavingsMsg'
Learn More

Methyl caprate, 98+%

Product Code. 10197183 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 10197183

Brand: Thermo Scientific Acros 385555000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 110-42-9 | C11H22O2 | 186.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-42-9
Molecular Formula C11H22O2
Molecular Weight (g/mol) 186.30
MDL Number MFCD00009580
InChI Key YRHYCMZPEVDGFQ-UHFFFAOYSA-N
Synonym methyl caprate, decanoic acid, methyl ester, methyl caprinate, capric acid methyl ester, decanoic acid methyl ester, methyl-n-caprate, uniphat a30, metholene 2095, methyl n-decanoate, methyl n-caprate
PubChem CID 8050
IUPAC Name methyl decanoate
SMILES CCCCCCCCCC(=O)OC

Specifications

Melting Point -18°C
Color Colorless
Density 0.8710g/mL
Boiling Point 226°C
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)8CO2CH3
Refractive Index 1.4240 to 1.4270 (20°C, 589nm)
Quantity 500 g
Specific Gravity 0.871
Solubility Information Solubility in water: practically insoluble
Formula Weight 186.29
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material Methyl caprate
Show More Show Less
compliance-icons
Product Identifier
  • Methyl caprate
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.