missing translation for 'onlineSavingsMsg'
Learn More

Methyl butyrate, 99%

Product Code. p-161170 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
100 g
250 g
Packaging:
Glass bottle
Unit Size:
100g
250g
5g
This item is not returnable. View return policy

Product Code. 10700872

Brand: Thermo Scientific Acros 414140050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 623-42-7 | C5H10O2 | 102.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 623-42-7
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.13
MDL Number MFCD00009391
InChI Key UUIQMZJEGPQKFD-UHFFFAOYSA-N
Synonym methyl butyrate, butanoic acid, methyl ester, methyl n-butyrate, butyric acid, methyl ester, methyl n-butanoate, butyric acid methyl ester, methyl-n-butyrate, n-butyric acid methyl ester, methyl butyrate natural, unii-cgx598508o
PubChem CID 12180
IUPAC Name methyl butanoate
SMILES CCCC(=O)OC

Specifications

Acidity 1mg KOH/g max.
Color Colorless to Yellow
Density 0.8970g/mL
Boiling Point 102°C
Flash Point 12°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)2COOCH3
Refractive Index 1.3850 to 1.39
Quantity 5 g
Merck Index 15, 6109
Specific Gravity 0.897
Solubility Information Solubility in water: insoluble
Formula Weight 102.13
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Methyl butyrate, 99%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.