missing translation for 'onlineSavingsMsg'
Learn More

Methyl butyrate, 99%

Product Code. 10015750 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
250 g
Packaging:
Glass bottle
Unit Size:
100g
250g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10015750 250 g Glass bottle 250g
10700872 5 g Glass bottle 5g
10701062 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10015750 Supplier Thermo Scientific Acros Supplier No. 414142500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 623-42-7 | C5H10O2 | 102.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 623-42-7
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.13
MDL Number MFCD00009391
InChI Key UUIQMZJEGPQKFD-UHFFFAOYSA-N
Synonym methyl butyrate, butanoic acid, methyl ester, methyl n-butyrate, butyric acid, methyl ester, methyl n-butanoate, butyric acid methyl ester, methyl-n-butyrate, n-butyric acid methyl ester, methyl butyrate natural, unii-cgx598508o
PubChem CID 12180
IUPAC Name methyl butanoate
SMILES CCCC(=O)OC

Specifications

Acidity 1mg KOH/g max.
Color Colorless to Yellow
Density 0.8970g/mL
Boiling Point 102°C
Flash Point 12°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)2COOCH3
Refractive Index 1.3850 to 1.39
Quantity 250 g
Merck Index 15, 6109
Specific Gravity 0.897
Solubility Information Solubility in water: insoluble
Formula Weight 102.13
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Methyl butyrate, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.