Learn More
Methyl beta-D-galactopyranoside
Description
It is a weak substrate and effective inducer of beta-D-galactosidase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is a weak substrate and effective inducer of beta-D-galactosidase.
Solubility
Soluble in water and methanol.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from heat. Keep it refrigerated.
Specifications
Specifications
| Melting Point | 175°C to 179°C |
| Quantity | 1 g |
| Beilstein | 81569 |
| Solubility Information | Soluble in water and methanol. |
| Optical Rotation | −16° (c=1.5 in Methanol) |
| Formula Weight | 194.18 |
| Chemical Name or Material | Methyl beta-D-galactopyranoside |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.