missing translation for 'onlineSavingsMsg'
Learn More

(Methyl benzoate)tricarbonylchromium, 98%, Thermo Scientific™

Product Code. 11418540
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11418540 1 g 1g
11408540 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11418540 Supplier Thermo Scientific Alfa Aesar Supplier No. 044368.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Catalyst for hydrogenation of unsaturated fats

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12125-87-0
Molecular Formula C11H8CrO5
Molecular Weight (g/mol) 272.18
MDL Number MFCD00009957
InChI Key GEALMZJLYGNHRA-UHFFFAOYSA-N
Synonym methyl benzoate tricarbonylchromium, tricarbonyl methyl benzoate chromium
PubChem CID 11780645
SMILES [Cr].[C-]#[O+].[C-]#[O+].[C-]#[O+].COC(=O)C1=CC=CC=C1

Specifications

Melting Point 95°C to 96°C
Color Orange
Quantity 1 g
UN Number UN3466
Sensitivity Air and moisture sensitive
IUPAC Name tris(methanidylidyneoxidanium) methyl benzoate chromium
Formula Weight 272.18
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material (Methyl benzoate)tricarbonylchromium

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.