missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ (Methyl benzoate)tricarbonylchromium, 98%

Catalyst for hydrogenation of unsaturated fats

Brand:  Alfa Aesar™ 044368.06

Code : NEW

Additional Details : CAS Number : 12125-87-0 Weight : 0.00500kg Transport : UN number : 3466 Chem class : 6.1 Pack group : II

Quantity 5g

 View more versions of this product

Product Code. 11408540

  • £255.90 / 5g


Estimated Shipment date
from Supplier 21-06-2018
Add to basket

Specifications

Chemical Identifiers

(Methyl benzoate)tricarbonylchromium
C11H8CrO5
MFCD00009957
(Methyl benzoate)chromium tricarbonyl
12125-87-0
272.18
UN3466

Specifications

(Methyl benzoate)tricarbonylchromium
Crystalline
C11H8CrO5
MFCD00009957
(Methyl benzoate)chromium tricarbonyl
95° to 96°C
Air & Moisture Sensitive
12125-87-0
98%
272.18
UN3466
Orange
5g
Videos
SDS