Learn More
Methyl benzoate, 99%
Description
Methyl benzoate is used in perfumery. It also finds use as a solvent. It acts as an intermediate and odor agents. Further, it is used to attract insects such as orchid bees. It is also used for cellulose esters, cellulose ethers, synthetic resin and rubber solvent and polyester fibers to assist in the preparation of flavor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Methyl benzoate is used in perfumery. It also finds use as a solvent. It acts as an intermediate and odor agents. Further, it is used to attract insects such as orchid bees. It is also used for cellulose esters, cellulose ethers, synthetic resin and rubber solvent and polyester fibers to assist in the preparation of flavor.
Solubility
Miscible with water.
Notes
Incompatible with strong oxidizing agents and strong bases.
Specifications
Specifications
| Melting Point | -12°C |
| Density | 1.094 |
| Boiling Point | 198°C to 200°C |
| Flash Point | 82°C (179°F) |
| Assay Percent Range | 99% |
| Refractive Index | 1.517 |
| Quantity | 5000 g |
| Beilstein | 1072099 |
| Merck Index | 14,6024 |
| Solubility Information | Miscible with water. |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.