missing translation for 'onlineSavingsMsg'
Learn More

Methyl Benzilate, TRC

Product Code. 30466794
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30466794 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30466794 Supplier Toronto Research Chemicals Supplier No. TRCM2897505G
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl Benzilate
CAS 76-89-1
Molecular Formula C15 H14 O3
Quantity 5 g
Synonym Benzilic Acid Methyl Ester, 2-Hydroxy-2,2-diphenylacetic Acid Methyl Ester, Methyl 2-Hydroxy-2,2-diphenylacetate, Methyl Diphenylglycolate, Methyl Hydroxydiphenylacetate, Methyl α-Hydroxydiphenylacetate, Methyl α-Phenylmandelate, NSC 57672, NSC 624
InChI Formula InChI=1S/C15H14O3/c1-18-14(16)15(17,12-8-4-2-5-9-12)13-10-6-3-7-11-13/h2-11,17H,1H3
SMILES COC(=O)C(O)(c1ccccc1)c2ccccc2
IUPAC Name methyl 2-hydroxy-2,2-diphenyl-acetate
Molecular Weight (g/mol) 242.27
Formula Weight 242.0943
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.