missing translation for 'onlineSavingsMsg'
Learn More

Methyl benzilate, 98%, Thermo Scientific Chemicals

Product Code. 11408436
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11408436 50 g 50g
11418436 250 g 250g
11498426 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11408436 Supplier Thermo Scientific Alfa Aesar Supplier No. A15907.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Store in cool dry place. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-89-1
Molecular Formula C15H14O3
Molecular Weight (g/mol) 242.274
MDL Number MFCD00004446
InChI Key LJFIHTFNTGQZJL-UHFFFAOYSA-N
Synonym methyl benzilate, methylbenzilate, methyl diphenylglycolate, benzilic acid methyl ester, benzilic acid, methyl ester, benzeneacetic acid, .alpha.-hydroxy-.alpha.-phenyl-, methyl ester, diphenylglycolic acid methyl ester, hydroxy-diphenyl-acetic acid methyl ester, methyl 2-hydroxy-2,2-di phenyl acetate, methyl benzillate
PubChem CID 66159
IUPAC Name methyl 2-hydroxy-2,2-diphenylacetate
SMILES COC(=O)C(C1=CC=CC=C1)(C2=CC=CC=C2)O

Specifications

Melting Point 73°C to 76°C
Boiling Point 187°C (13 mmHg)
Quantity 50 g
Beilstein 2121492
Merck Index 14,1080
Solubility Information Soluble in methanol. Insoluble in water.
Formula Weight 242.28
Percent Purity 98%
Chemical Name or Material Methyl benzilate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.