missing translation for 'onlineSavingsMsg'
Learn More

Methyl benzenesulfonate, 99%

Product Code. 10095680 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10095680 100 g Glass bottle 100g
10732004 25 g Glass bottle 25g
11924571 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10095680 Supplier Thermo Scientific Acros Supplier No. 413991000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 80-18-2 | C7H8O3S | 172.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-18-2
Molecular Formula C7H8O3S
Molecular Weight (g/mol) 172.20
MDL Number MFCD00014737
InChI Key CZXGXYBOQYQXQD-UHFFFAOYSA-N
Synonym benzenesulfonic acid, methyl ester, methyl benzenesulphonate, methyl besylate, methylbenzenesulfonate, ccris 9156, benzenesulfonic acid methyl ester, methyl benzene sulfonate, methyl ester of benzenesulphonic acid, methylbenzene sulfonate, pubchem22730
PubChem CID 6630
IUPAC Name methyl benzenesulfonate
SMILES COS(=O)(=O)C1=CC=CC=C1

Specifications

Melting Point -4.0°C
Color Colorless to Yellow
Density 1.3000g/mL
Boiling Point 280.0°C
Flash Point 143°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Refractive Index 1.5140 to 1.5160
Quantity 100 g
Beilstein 07,645
Specific Gravity 1.3
Formula Weight 172.2
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Methyl benzenesulfonate, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Methyl benzenesulfonate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.