missing translation for 'onlineSavingsMsg'
Learn More

Methyl benzenesulfinate, 98%

Product Code. 11925761 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
Unit Size:
25mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11925761 5 mL 5mL
11935761 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 11925761 Leverantör Thermo Scientific Acros Leverantörsnummer 438610050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

CAS: 670-98-4 | C7H8O2S | 156.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 670-98-4
Molecular Formula C7H8O2S
Molecular Weight (g/mol) 156.2
InChI Key PSNSVDSRLUYDKF-UHFFFAOYSA-N
Synonym methylbenzenesulfinate, benzenesulfinic acid methyl ester, methyl phenylsulfinate, methyl benzene sulfinate, benzenesulfinic acid,methyl ester, benzenesulfinic acid, methyl ester, r, benzenesulfinic acid, methyl ester, s, methoxy phenylsulfinyl, acmc-20mem2, acmc-20mg8f
PubChem CID 2759784
IUPAC Name methyl benzenesulfinate
SMILES COS(=O)C1=CC=CC=C1

Specifikationer

Color Colorless to Yellow
Density 1.1940g/mL
Boiling Point 110.0°C to 112.0°C (16.0 Torr)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 5 mL
Specific Gravity 1.194
Formula Weight 156.2
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Methyl benzenesulfinate
Visa mer Visa mindre
Produktbeteckning
  • Methyl benzenesulfinate, 98%

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.