missing translation for 'onlineSavingsMsg'
Learn More

Methyl Anthranilate, TRC

Product Code. 30473183
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
Product Code. Quantity unitSize
30473183 100 g 100g
1 options
This item is not returnable. View return policy

Product Code. 30473183

Brand: Toronto Research Chemicals TRCM265070100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Anthranilic Acid Methyl Ester
CAS 134-20-3
Molecular Formula C8 H9 N O2
Quantity 100 g
Synonym Anthranilic acid','methyl ester (6CI,8CI), 2-(Methoxycarbonyl)aniline, 2-Aminobenzoic acid methyl ester, 2-Carbomethoxyaniline, Bird Shield, Fog Force, Grain 96-1, Methyl 2-aminobenzoate, Methyl 6-aminobenzoate, Methyl anthranilate, Methyl o-aminobenzoate, NSC 3109, ReJex-iT, RejeX-it Fog Force, Rejex-iT AP 50, Rejex-iT TP 40, Sunarome UVA, [2-(Methoxycarbonyl)phenyl]amine, o-(Methoxycarbonyl)aniline, o-Aminobenzoic acid methyl ester, o-Carbomethoxyaniline
InChI Formula InChI=1S/C8H9NO2/c1-11-8(10)6-4-2-3-5-7(6)9/h2-5H,9H2,1H3
SMILES COC(=O)c1ccccc1N
IUPAC Name methyl 2-aminobenzoate
Molecular Weight (g/mol) 151.16
Formula Weight 151.0633
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.