missing translation for 'onlineSavingsMsg'
Learn More

Methyl α-aminoisobutyrate hydrochloride, 98%, Thermo Scientific™

Product Code. 10346774 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10346774 5 g Glass bottle 5g
10634165 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10346774 Supplier Thermo Scientific Acros Supplier No. 397980050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemische Identifikatoren

CAS 15028-41-8
MDL Number MFCD00214247
InChI Key NVWZNEDLYYLQJC-UHFFFAOYSA-N
Synonym methyl 2-amino-2-methylpropanoate hydrochloride, methyl 2-aminoisobutyrate hydrochloride, h-aib-ome.hcl, alpha-aminoisobutyric acid methyl ester hydrochloride, 2-aminoisobutyric acid methyl ester hydrochloride, methyl alpha-aminoisobutyrate hydrochloride, methyl 2-methylalaninate hydrochloride, alanine, 2-methyl-, methyl ester, hydrochloride, aib-ome hydrochloride, pubchem16472
PubChem CID 13258034
IUPAC Name methyl 2-amino-2-methylpropanoate;hydrochloride
SMILES CC(C)(C(=O)OC)N.Cl

Spezifikation

Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C5H12ClNO2
Quantity 5 g
Molecular Weight (g/mol) 153.606
Formula Weight 151.64
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material Methyleα-aminoisobutyrate hydrochloride
Mehr anzeigen Weniger anzeigen
compliance-icons
Produktidentifikator
  • Methyl alpha-aminoisobutyrate hydrochloride
Signalwort
  • Warning
Gefahrenkategorie
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Gefahrenhinweis
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Sicherheitshinweis
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.